Potassium iodide 30% in aqueous solution analytical reagent
Supplier: Bernd Kraft
Jump to:
About this item
Synonyms: Dipotassium diiodide, Kali iodide
- CAS Number:7681-11-0
- Formula:Kl
- MDL Number:MFCD00011405
- Molecular Weight:166 g/mol
- Storage Temperature:Ambient
- EINECS:231-659-4
5 Options Available Below
Product Details & Documents
Synonyms: Dipotassium diiodide, Kali iodide
- CAS Number:7681-11-0
- Formula:Kl
- MDL Number:MFCD00011405
- Molecular Weight:166 g/mol
- Storage Temperature:Ambient
- EINECS:231-659-4
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
This product has been blocked by your organization. Please contact your purchasing department for more information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



