Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide 20% in aqueous solution
Potassium iodide 20% in aqueous solution
Catalog # HACH1456842
Supplier:  Hach
CAS Number:  
Potassium iodide 20% in aqueous solution
Catalog # HACH1456842
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :166 g/mol
Storage Temperature:Ambient
EINECS:231-659-4
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405