
Sulphuric acid 40% for determination of gas metabolism acc. to Knipping
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Boiling Pt:∼100 °C (1013 hPa)
Density:1.1…1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
Frequently Bought Together