Order Entry
Belgium
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Corrosive
Environment
Zinc sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09991.0001
Supplier:  Merck
CAS Number:  
Corrosive
Environment
Zinc sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Catalog # 1.09991.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.16 g/cm3 (20 °C)
  • pH:
    2.2 (H2O, 20 °C)
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

About this item

 :
 :161.45 g/mol
ADR:9,
 :M-Clarity™ quality level = MQ100
EINECS:231-793-3
UN:3082
 :GHS05
 :GHS09
Storage Temperature:Ambient
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302