Order Entry
Belgium
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # ROTHP723.1
Supplier:  Roth Carl
CAS Number:  
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # ROTHP723.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302