Order Entry
Belgium
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30498.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Environment
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30498.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l

Specification Test Results

About this item

 :
 :161.45 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302