Order Entry
Belgium
ContactUsLinkComponent
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Irritant
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Catalog # 1.08879.1000
Supplier:  Merck
CAS Number:  
Irritant
Zinc sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Catalog # 1.08879.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0995 - 0.1005
  • Measurement uncertainty:
    +/- 0.0003
  • Traceability:
    NIST SRM

Specification Test Results

About this item

 :
 :161.45 g/mol
ADR:9,III
 :M-Clarity™ quality level = MQ100
EINECS:231-793-3
UN:3082
 :GHS07
Storage Temperature:Ambient
CAS Number:7733-02-0
Formula:O₄SZn
MDL Number:MFCD00011302