Silver sulphate 80 g/l in sulphuric acid for COD determination, Supelco®
Catalog # 1.01574.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Concentration (Ag₂SO₄):78.8 - 81.2 g/l
- Concentration (H₂SO₄):1665 - 1735 g/l
Specification Test Results
About this item
Synonyms: Disilver sulphate
:
:311.8 g/mol
EINECS:233-653-7
UN:3264
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407
Boiling Pt:1085
Melting Pt:655
ADR:8 II
Merck Index:13,08603
:M-Clarity™ quality level = MQ100