Order Entry
Belgium
ContactUsLinkComponent
Silver sulphate 26,6 g/l in sulphuric acid
Silver sulphate 26,6 g/l in sulphuric acid
Catalog # KRAF05378.3700
Supplier:  Bernd Kraft
CAS Number:  
undefined
Silver sulphate 26,6 g/l in sulphuric acid
Catalog # KRAF05378.3700
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Disilver sulphate
 :
 :311.8 g/mol
EINECS:233-653-7
UN:1830
ADR:8,II
 :GHS05
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407