Order Entry
Belgium
ContactUsLinkComponent
 
Silver sulphate 10 g/l in sulphuric acid
  CHMBCL02.2603.2500
 :  CHEMLAB
 :  
undefined
Silver sulphate 10 g/l in sulphuric acid
  CHMBCL02.2603.2500
 :  CHEMLAB
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

 

 

 : Disilver sulphate
 :
 :311.8 g/mol
 :233-653-7
 :1830
 :GHS05
 :10294-26-5
 :Ag₂SO₄
 :MFCD00003407
 :1085
 :655
 :8 II
 :13,08603