Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Ammonium cerium(IV) nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30483.295
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Ammonium cerium(IV) nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 30483.295
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :548.22 g/mol
EINECS:240-827-6
 :GHS07
CAS Number:16774-21-3
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121