ContactUsLinkComponent
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Corrosive
Environment
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Supplier:  Merck
Certificates
Corrosive
Environment
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, Titripur® Reag. Ph. Eur. volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.0995 - 0.1005
  • Measurement uncertainty:
    +/- 0.0003
  • Traceability:
    NIST SRM