Iodine 0.05 mol/l (0.1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
UN:3264
ADR:8,II
Specification Test Results
- Titer (N):0.0997 - 0.1003
Specification Test Results
Frequently Bought Together