Ammonium iron(II) sulphate 0.06 mol/l (0.06 N) in aqueous solution for COD determination
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:10045-89-3
:
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039
:284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
Frequently Bought Together