Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Supplier: MARKET SOURCE PART PROCESS
Certificates
About this item
CAS Number:10045-89-3
:
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039
:284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
Frequently Bought Together