Order Entry
Belgium
ContactUsLinkComponent
Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Catalog # 30882.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium iron(II) sulphate 0.05 mol/l (0.05 N) in aqueous solution
Catalog # 30882.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0499 - 0.0501 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039