Order Entry
Belgium
ContactUsLinkComponent
 
Ammonium iron(II) sulphate 0.025 mol/l (0.025 N) in aqueous solution
Catalog # MSPP80010697.1000
CAS Number:  
Ammonium iron(II) sulphate 0.025 mol/l (0.025 N) in aqueous solution
Catalog # MSPP80010697.1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate
 :
 :284.05 g/mol
Storage Temperature:Ambient
EINECS:233-151-8
CAS Number:10045-89-3
Formula:H₈FeN₂O₈S₂
MDL Number:MFCD00051039