Order Entry
Belgium
ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) Reag. Ph. Eur. Chapter 4 (4.2.2)
Flammable
Corrosive
Irritant
Environment
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) Reag. Ph. Eur. Chapter 4 (4.2.2)
Catalog # REGE3008300
Supplier:  REAGECON
CAS Number:  
Flammable
Corrosive
Irritant
Environment
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) Reag. Ph. Eur. Chapter 4 (4.2.2)
Catalog # REGE3008300
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:3286
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS09
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425