Order Entry
Belgium
ContactUsLinkComponent
Magnesium chloride 0.025 mol/l (0.05 N) in aqueous solution
Magnesium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # ADVRAB-0359
CAS Number:  
Magnesium chloride 0.025 mol/l (0.05 N) in aqueous solution
Catalog # ADVRAB-0359
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106