
Barium chloride 0.1 mol/l (0.2 N) in aqueous solution
Catalog # KRAF05592.6020
Supplier: Bernd Kraft
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:208.24 g/mol
Density:1.034 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
:GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445