Order Entry
Belgium
ContactUsLinkComponent
Barium chloride 0.05 mol/l (0.1 N) in aqueous solution
Irritant
Barium chloride 0.05 mol/l (0.1 N) in aqueous solution
Catalog # KRAF03152.3000
Supplier:  Bernd Kraft
CAS Number:  
Irritant
Barium chloride 0.05 mol/l (0.1 N) in aqueous solution
Catalog # KRAF03152.3000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :208.24 g/mol
Density:1.034 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:3287
ADR:6.1,III
 :GHS07
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445