

Hydrochloric acid 36%, VLSI Selectipur® for the electronics industry
Catalog # ELCH51153627_U
Supplier: BASF ELECTRONIC CHEMICALS
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
Melting Pt:-30 °C
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
REACH:01-2119484862-27
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324