
Sodium hydroxyde 1 mol/l, EMPROVE® EXPERT, SAFC®
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:1,04 g/cm³ (25 °C)
:Température ambiante
:215-185-5
:1824
:8,
:GHS05
- Assay (Acidimetric, NaOH):0.99 - 1.01 mol/l
- Carbonate (as Na₂CO₃):≤1.0%
- Chloride (Cl):≤5 ppm
- Sulfate (SO₄):≤30 ppm
- Heavy Metals (as Pb):≤5.0 ppm
- Al (Aluminium):≤10 ppm
- Fe (Iron):≤3 ppm
- Residual Solvents (ICH Q3C):Excluded by Production Process
- Endotoxins:≤2.0 I.U./ml
- Total Aerobic Microbial Count (TAMC):≤100 CFU/g
- Total Combined Yeasts/Moulds Count (TYMC):≤10 CFU/g
Frequently Bought Together