Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # 1.11787.2500
Supplier:  Merck
CAS Number:  
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 2 mol/l (2 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # 1.11787.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :56.11 g/mol
ADR:3 (6.1, 8),II
 :M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:3286
 :GHS02
 :GHS06
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553