ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Flammable
Toxic
Corrosive
Health
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Supplier:  Merck
Certificates
Specification Test Results
  • Form:
    liquid
  • Amount-of-substance concentration:
    0.4975 - 0.5025
  • Measurement uncertainty:
    +/- 0.0015
  • Traceability:
    NIST SRM