

Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Melting Pt:–114.5 °C (EtOH)
Density:0.85 g/cm³ (20 °C)
Flash Pt:12 °C (EtOH)
Storage Temperature:Ambient
EINECS:215-181-3
ADR:3 (8),II
:M-Clarity™ quality level = MQ100
UN:2924
:GHS02
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.4975 - 0.5025
- Measurement uncertainty:+/- 0.0015
- Traceability:NIST SRM
Specification Test Results
Frequently Bought Together