Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Flammable
Corrosive
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7131.1000
Supplier:  Avantor
CAS Number:  
Flammable
Corrosive
Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7131.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (mol/l):
    0.4975 - 0.5025

Specification Test Results

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:2924
ADR:3,II
 :GHS02
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553