Order Entry
Belgium
ContactUsLinkComponent
 
 
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM volumetric solution
  31950.296
 :  
undefined
 
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, AVS TITRINORM volumetric solution
  31950.296
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.499 - 0.501 mol/l

 

 

 :
 :56.11 g/mol
 :1.023 g/cm³ (20 °C)
 :Ambient
 :215-181-3
 :1814
 :8,II
 :GHS05
 :1310-58-3
 :KOH
 :MFCD00003553