Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.11586.5000
Supplier:  Merck
CAS Number:  
Potassium hydroxide 0.5 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 1.11586.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :56.11 g/mol
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:1814
 :GHS05
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553