


Potassium hydroxide 0.1 mol/l (0.1 N) in methanol, Titripur® volumetric solution, Supelco®
Catalog # 1.11587.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:56.11 g/mol
ADR:3 (6.1, 8),II
:M-Clarity™ quality level = MQ100
EINECS:215-181-3
UN:3286
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553