Order Entry
Belgium
ContactUsLinkComponent
 
 
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, AVS TITRINORM volumetric solution
  31303.291
 :  
undefined
 
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, AVS TITRINORM volumetric solution
  31303.291
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • NIST traceable:
    Confirmed
  • Appearance:
    Clear colourless liquid
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l

 

 

 :
 :56.11 g/mol
 :Ambient
 :215-181-3
 :2924
 :3 (8),II
 :GHS02
 :1310-58-3
 :KOH
 :MFCD00003553