Order Entry
Belgium
ContactUsLinkComponent
 
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
  VWRS5949.9020
 :  
undefined
Sodium dodecyl sulphate (SDS) 100 g/l in aqueous solution
  VWRS5949.9020
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    20 L
  • Seq:
    5302

 

 

 :
 :288.38 g/mol
 :1…1.1 g/cm³ (20 °C)
 :Ambient
 :205-788-1
 :151-21-3
 :CH₃(CH₂)₁₁OSO₃Na
 :MFCD00036175