Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 2 mol/l (4 N) in aqueous solution
Sulphuric acid 2 mol/l (4 N) in aqueous solution
Catalog # CHMBCL05.2619.5000
Supplier:  CHEMLAB
CAS Number:  
Sulphuric acid 2 mol/l (4 N) in aqueous solution
Catalog # CHMBCL05.2619.5000
Supplier:  CHEMLAB
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

About this item

 :
 :98.08 g/mol
Boiling Pt:330
EINECS:231-639-5
UN:2796
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.146 g/cm³ (20 °C)
ADR:8 II