
Sulphuric acid 2.5 mol/l (5 N) in aqueous solution volumetric solution
Supplier: REAGECON
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.146 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
Frequently Bought Together