Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 2.5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®
Corrosive
Sulphuric acid 2.5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09912.0001
Supplier:  Merck
CAS Number:  
Corrosive
Sulphuric acid 2.5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®
Catalog # 1.09912.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Boiling Point:
    103 °C
  • Density:
    1.15 g/cm3 (20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.146 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:231-639-5
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589