Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:13,04497
Melting Pt:18 °C
Density:1.227 g/cm³
Flash Pt:160 °C
Storage Temperature:Ambient
EINECS:200-289-5
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
Specifications
Specification Test Results
- Appearance:Clear colorless to almost colorless Viscous liquid
- Refractive index:1.4490 to 1.4550 (20°C, 589 nm)
- Assay:83.5 to 88.5 %
- Water:12 to 16 %
Specification Test Results
Frequently Bought Together