

2-Propanol 70% in water
Catalog # 563935-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:69 - 71 %
Specification Test Results
About this item
:
:60.1 g/mol
Boiling Pt:81…83 °C
Density:0.858 g/cm³ (20 °C)
Flash Pt:22 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674