Sulphuric acid 0.005 mol/l (0.01 N) in aqueous solution for HPLC
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:1.01 g/cm³ (20 °C)
:Ambient
:231-639-5
:2796
:8,II
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals