Order Entry
Belgium
ContactUsLinkComponent
 
 
Hydrochloric acid 0.2 M HCl (0.2N) ≥97% volumetric solution, Fluka™
 
 
Hydrochloric acid 0.2 M HCl (0.2N) ≥97% volumetric solution, Fluka™

 

  • Appearance (trubidity):
    complying
  • Appearance (color):
    complying
  • Appearance (form):
    complying
  • Traceable to NIST SRM:
    effective
  • Assay:
    0.1900 - 0.2100 N

 

 

Frequently Bought Together