Order Entry
Belgium
ContactUsLinkComponent
 
Hydrochloric acid 0.01 mol/l (0.01 N), VOLUSOL® volumetric solution
  311341.1000
 :  
undefined
Hydrochloric acid 0.01 mol/l (0.01 N), VOLUSOL® volumetric solution
  311341.1000
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Titer (20°C; real value 0.2 % accuracy):
    0.00998 - 0.01002 mol/l
  • NIST traceable:
    Confirmed

 

 

 :
 :1 g/cm³ (20 °C)
 :Ambient
 :231-595-7
 :8,III
 :36.46 g/mol
 :1789
 :7647-01-0
 :HCl
 :MFCD00011324