Order Entry
Belgium
ContactUsLinkComponent
Guanidinium hydrochloride 8 mol/l in aqueous solution
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution
Catalog # A0860.0500
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution
Catalog # A0860.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :95.53 g/mol
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026