Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution, Biotechnology Grade
Catalog # M113-4L
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Guanidinium hydrochloride 8 mol/l in aqueous solution, Biotechnology Grade
Catalog # M113-4L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Protease:
    none detected

Specification Test Results

About this item

 :
 :95.53 g/mol
EINECS:200-002-3
 :GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026