EDTA disodium salt 0.0178 mol/l (N/28) in water, AVS TITRINORM for hardness of water determinations
Catalog # 30029.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20 °C; real value 0,2 % accuracy):0,017764 - 0,01784 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:336.21 g/mol
Density:1.001 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672