Order Entry
Belgium
ContactUsLinkComponent
Glutaraldehyde 25% in aqueous solution
Toxic
Corrosive
Health
Environment
Glutaraldehyde 25% in aqueous solution
Catalog # ROTH4157.1
Supplier:  Roth Carl
CAS Number:  
Toxic
Corrosive
Health
Environment
Glutaraldehyde 25% in aqueous solution
Catalog # ROTH4157.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
 :100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:>71 °C
Storage Temperature:Refrigerator
EINECS:203-856-5
UN:2927
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:111-30-8
Formula:C₅H₈O₂
MDL Number:MFCD00007025