Acetic acid 1 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Frequently Bought Together
Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Pasteur pipettes, disposable
: Brand
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
: MERCK PRODUCTION CHEMICALS
VWR®, Nitrile Gloves
: VWR Collection
Test tubes, Corning® Gosselin™
: Corning
Hydrochloric acid 1 mol/l (1N) volumetric solution
: PanReac AppliChem