Order Entry
Belgium
ContactUsLinkComponent
Sodium pyruvate 0.1 mol/l in aqueous solution
Sodium pyruvate 0.1 mol/l in aqueous solution
Catalog # HYCLSH30239.01
CAS Number:  
undefined
Sodium pyruvate 0.1 mol/l in aqueous solution
Catalog # HYCLSH30239.01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Seq:
    2440

Specifications

About this item

 :
 :110.04 g/mol
Storage Temperature:Ambient
EINECS:204-024-4
CAS Number:113-24-6
Formula:CH₃COCOONa
MDL Number:MFCD00002586