Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur., Supelco®
Catalog # 1.59712.0010
Supplier:  Merck
CAS Number:  
Corrosive
(±)-α-Methoxyphenylacetic acid Reag. Ph. Eur., Supelco®
Catalog # 1.59712.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :166.18 g/mol
Boiling Pt:165 °C (24 hPa)
Melting Pt:69…71 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:230-300-9
 :GHS05
CAS Number:7021-09-2
Formula:C₉H₁₀O₃
MDL Number:MFCD00064217