N-Acetyl-DL-tryptophan, EMPROVE® EXPERT BP, Ph. Eur., extra pure for biopharmaceutical production low in endotoxins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Ac-DL-Trp-OH, N-Acetyl-DL-tryptophan
CAS Number:87-32-1
:
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD00005644
:246.27 g/mol
Melting Pt:205…206 °C
Storage Temperature:Ambient
EINECS:201-739-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together