

(±)-1-Methoxy-2-propanol
Supplier: Roth Carl
Certificates
About this item
CAS Number:107-98-2
:
Formula:CH₃CH(OH)CH₂OCH₃
MDL Number:MFCD00004537
Boiling Pt:119.6 °C (1013 hPa)
Melting Pt:–97 °C
Density:0.92 g/cm³ (20 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:203-539-1
ADR:3,III
:90.12 g/mol
UN:3092
:GHS02
:GHS07
Frequently Bought Together