Order Entry
Belgium
ContactUsLinkComponent
Diethyl ether, dehydrated (max. 0.01% H₂O) ≥99.5% (by GC, corrected for water content) stabilised for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Diethyl ether, dehydrated (max. 0.01% H₂O) ≥99.5% (by GC, corrected for water content) stabilised for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9237-03
Supplier:  Avantor
CAS Number:  
undefined
Diethyl ether, dehydrated (max. 0.01% H₂O) ≥99.5% (by GC, corrected for water content) stabilised for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9237-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:
    ≤0.02
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:
    ≤0.07
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 231 nm:
    ≤0.4
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤220
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.5 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb
  • Assay (by GC, corrected for water, exclusive of preservative):
    ≥99.5 %
  • Peroxide (as H₂O₂):
    ≤5 ppm
  • Preservative (C₂H₅OH):
    1.5 - 3.5 %
  • Residue after Evaporation:
    ≤5 ppm
  • Titrable Acid (µeq/g):
    ≤0.2
  • Water (H₂O)(by coulometry):
    ≤0.01 %

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
 :Stabilised with ethanol 1.5 - 3.5%
REACH:01-2119535785-29
 :GHS02
 :GHS07
 :GHS08
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646